1-(4-Bromo-5-methyl-2-thienyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

270497-1G 270497-500MG 270497-5G 270497-25G
77467-902EA 197.76 USD
77467-902 77480-598 77490-940 77500-996
1-(4-Bromo-5-methyl-2-thienyl)-1-propanone ≥95%
1-(4-Bromo-5-methyl-2-thienyl)-1-propanone

Precaución: For research use only

Formula: C₈H₉BrOS
MW: 233.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860028
CAS Number: 887576-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR