1-Ethoxy-3-(ethylthio)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

270225-1G 270225-500MG 270225-5G 270225-25G
77467-898EA 415.09 USD
77467-898 77480-594 77490-936 77500-992
1-Ethoxy-3-(ethylthio)-2-methylbenzene ≥95%
1-Ethoxy-3-(ethylthio)-2-methylbenzene

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34878937
CAS Number: 85428-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR