1-(Ethylthio)-2-fluoro-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

270139-1G 270139-500MG 270139-5G 270139-25G
77467-894EA 415.09 USD
77467-894 77480-590 77490-932 77500-988
1-(Ethylthio)-2-fluoro-3-(trifluoromethyl)benzene ≥95%
1-(Ethylthio)-2-fluoro-3-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₉H₈F₄S
MW: 224.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22484166
CAS Number: 1824659-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR