3-Fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

269948-1G 269948-500MG 269948-5G 269948-25G
77467-890EA 415.09 USD
77467-890 77480-586 77490-928 77500-984
3-Fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol ≥95%
3-Fluoro-α-methyl-α-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068012
CAS Number: 1157636-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR