3-Bromo-α-methyl-α-(2-methylpropyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

269903-1G 269903-500MG 269903-5G 269903-25G
77467-886EA 415.09 USD
77467-886 77480-582 77490-924 77500-980
3-Bromo-α-methyl-α-(2-methylpropyl)benzenemethanol ≥97%
3-Bromo-α-methyl-α-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
MW: 257.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068100
CAS Number: 1157637-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR