α-Ethyl-α-methyl-4-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

269864-1G 269864-500MG 269864-5G
77467-882EA 648.9 USD
77467-882 77480-578 77490-920
α-Ethyl-α-methyl-4-(2-methylpropyl)benzenemethanol ≥95%
α-Ethyl-α-methyl-4-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
MW: 206.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526619
CAS Number: 1443326-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR