5-Bromo-4-iodo-1,3-benzodioxole ≥95%

Supplier: AOBChem USA

Warning

26947-1G 26947-5G 26947-25G
77467-878EA 60.77 USD
77467-878 77490-916 77500-974
5-Bromo-4-iodo-1,3-benzodioxole ≥95%
5-Bromo-4-iodo-1,3-benzodioxole

Precaución: For research use only

Formula: C₇H₄BrIO₂
Storage Temperature: Ambient
MDL Number: MFCD27756694
CAS Number: 1300118-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR