3-(2-Fluoro-5-(trifluoromethyl)phenyl)tetrahydrofuran-3-ol ≥95%

Supplier: AOBChem USA

Warning

269399-1G 269399-500MG 269399-5G 269399-25G
77467-874EA 415.09 USD
77467-874 77480-572 77490-912 77500-972
3-(2-Fluoro-5-(trifluoromethyl)phenyl)tetrahydrofuran-3-ol ≥95%
3-(2-Fluoro-5-(trifluoromethyl)phenyl)tetrahydrofuran-3-ol

Precaución: For research use only

Formula: C₁₁H₁₀F₄O₂
MW: 250.1895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34878640
CAS Number: 2969319-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR