5-Chloro-2-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

26907-1G 26907-500MG 26907-5G 26907-25G
77467-866EA 195.7 USD
77467-866 77480-564 77490-906 77500-966
5-Chloro-2-(methylsulfanyl)benzoic acid ≥97%
5-Chloro-2-(methylsulfanyl)benzoic acid

Precaución: For research use only

Formula: C₈H₇ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD04119794
CAS Number: 62176-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR