3-(3-Fluorophenyl)tetrahydro-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268493-1G 268493-500MG 268493-5G
77467-854EA 447.02 USD
77467-854 77480-554 77490-894
3-(3-Fluorophenyl)tetrahydro-3-furanol ≥95%
3-(3-Fluorophenyl)tetrahydro-3-furanol

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18332720
CAS Number: 1339412-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR