α-(4-Methoxyphenyl)benzeneethanol ≥95%

Supplier: AOBChem USA

Warning

268168-1G 268168-500MG 268168-5G 268168-25G
77467-842EA 415.09 USD
77467-842 77480-542 77490-882 77500-946
α-(4-Methoxyphenyl)benzeneethanol ≥95%
α-(4-Methoxyphenyl)benzeneethanol

Precaución: For research use only

Formula: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00155072
CAS Number: 5422-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR