5-Bromo-α-butyl-2-fluoro-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

268122-1G 268122-500MG 268122-5G 268122-25G
77467-838EA 390.37 USD
77467-838 77480-538 77490-878 77500-942
5-Bromo-α-butyl-2-fluoro-α-methylbenzenemethanol ≥95%
5-Bromo-α-butyl-2-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
MW: 275.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34878000
CAS Number: 2003391-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR