α-Butyl-2-fluoro-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

268071-1G 268071-500MG 268071-5G 268071-25G
77467-834EA 260.59 USD
77467-834 77480-534 77490-874 77500-938
α-Butyl-2-fluoro-α-methylbenzenemethanol ≥95%
α-Butyl-2-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21128216
CAS Number: 1489581-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR