3-Cyclohexen-1-ol, 4-bromo-, 1-benzoate ≥95%

Supplier: AOBChem USA

Warning

26752-1G 26752-500MG 26752-5G
77467-830EA 830.18 USD
77467-830 77480-530 77490-870
3-Cyclohexen-1-ol, 4-bromo-, 1-benzoate ≥95%
3-Cyclohexen-1-ol, 4-bromo-, 1-benzoate

Precaución: For research use only

Formula: C₁₃H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD27756674
CAS Number: 1450812-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR