(3-Bromo-2-fluoro-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

26714-1G 26714-500MG 26714-5G
77467-826EA 687.01 USD
77467-826 77480-526 77490-866
(3-Bromo-2-fluoro-4-methoxyphenyl)methanol ≥97%
(3-Bromo-2-fluoro-4-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27959301
CAS Number: 1155877-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR