1-(3-Bromo-2-fluorophenyl)-1,2-ethanediol ≥95%

Supplier: AOBChem USA

Warning

267028-1G 267028-500MG 267028-5G 267028-25G
77467-822EA 423.33 USD
77467-822 77480-522 77490-862 77500-932
1-(3-Bromo-2-fluorophenyl)-1,2-ethanediol ≥95%
1-(3-Bromo-2-fluorophenyl)-1,2-ethanediol

Precaución: For research use only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
CAS Number: 1866646-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR