1-(3-Bromophenyl)-1,2-ethanediol ≥95%

Supplier: AOBChem USA

Warning

266880-1G 266880-500MG 266880-5G 266880-25G
77467-818EA 355.35 USD
77467-818 77480-518 77490-858 77500-930
1-(3-Bromophenyl)-1,2-ethanediol ≥95%
1-(3-Bromophenyl)-1,2-ethanediol

Precaución: For research use only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
CAS Number: 402937-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR