2-(2,3-Difluorophenyl)tetrahydro-2H-pyran-2-ol ≥95%

Supplier: AOBChem USA

Warning

266848-1G 266848-500MG 266848-5G 266848-25G
77467-814EA 415.09 USD
77467-814 77480-514 77490-854 77500-926
2-(2,3-Difluorophenyl)tetrahydro-2H-pyran-2-ol ≥95%
2-(2,3-Difluorophenyl)tetrahydro-2H-pyran-2-ol

Precaución: For research use only

Formula: C₁₁H₁₂F₂O₂
MW: 214.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34877797
CAS Number: 2969352-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR