2-Iodo-5-methylbenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

266828-1G 266828-500MG 266828-5G 266828-25G
77467-810EA 415.09 USD
77467-810 77480-510 77490-850 77500-920
2-Iodo-5-methylbenzo[b]thiophene ≥95%
2-Iodo-5-methylbenzo[b]thiophene

Precaución: For research use only

Formula: C₉H₇IS
MW: 274.1213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34877794
CAS Number: 2930828-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR