1-[4'-(1-Methylethoxy)[1,1'-biphenyl]-4-yl]ethanone ≥95%

Supplier: AOBChem USA

Warning

266559-1G 266559-500MG 266559-5G 266559-25G
77467-806EA 415.09 USD
77467-806 77480-506 77490-846 77500-916
1-[4'-(1-Methylethoxy)[1,1'-biphenyl]-4-yl]ethanone ≥95%
1-[4'-(1-Methylethoxy)[1,1'-biphenyl]-4-yl]ethanone

Precaución: For research use only

Formula: C₁₇H₁₈O₂
MW: 254.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11922908
CAS Number: 1176477-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR