1-(3,5-Dimethyl-4-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

266478-1G 266478-500MG 266478-5G 266478-25G
77467-802EA 341.96 USD
77467-802 77480-502 77490-842 77500-914
1-(3,5-Dimethyl-4-propoxyphenyl)ethanone ≥95%
1-(3,5-Dimethyl-4-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD13813474
CAS Number: 1119425-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR