3-Bromo-2-fluoro-6-(trifluoromethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

265986-1G 265986-500MG 265986-5G
77467-798EA 691.13 USD
77467-798 77480-498 77490-838
3-Bromo-2-fluoro-6-(trifluoromethoxy)benzaldehyde ≥95%
3-Bromo-2-fluoro-6-(trifluoromethoxy)benzaldehyde

Precaución: For research use only

Formula: C₈H₃BrF₄O₂
MW: 287.01 g/mol
Storage Temperature: Ambient
CAS Number: 2168738-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR