tert-Butyl 3-(3-chloro-2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

264887-1G 264887-500MG 264887-5G 264887-25G
77467-786EA 415.09 USD
77467-786 77480-486 77490-826 77500-900
tert-Butyl 3-(3-chloro-2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%
tert-Butyl 3-(3-chloro-2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate

Precaución: For research use only

Formula: C₁₄H₁₇ClFNO₃
MW: 301.7410 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34877176
CAS Number: 2969380-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR