2-Bromo-6-chloropyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

26471-1G 26471-500MG 26471-5G 26471-25G
77467-782EA 99.91 USD
77467-782 77480-482 77490-822 77500-896
2-Bromo-6-chloropyridin-3-ol ≥97%
2-Bromo-6-chloropyridin-3-ol

Precaución: For research use only

Formula: C₅H₃BrClNO
MW: 208.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972190
CAS Number: 1020253-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR