1-(2,3-Dimethylphenyl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264566-1G 264566-500MG 264566-5G 264566-25G
77467-778EA 315.18 USD
77467-778 77480-478 77490-816 77500-890
1-(2,3-Dimethylphenyl)-2-methyl-1-propanone ≥95%
1-(2,3-Dimethylphenyl)-2-methyl-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16301701
CAS Number: 1226148-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR