1-[3-(1,1-Dimethylethyl)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264516-1G 264516-500MG 264516-5G
77467-774EA 668.47 USD
77467-774 77480-474 77490-812
1-[3-(1,1-Dimethylethyl)phenyl]-1-propanone ≥95%
1-[3-(1,1-Dimethylethyl)phenyl]-1-propanone

Precaución: For research use only

Formula: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD11617657
CAS Number: 1228926-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR