Ethyl 2,6-dibromopyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2,6-dibromoisonicotinate

26380-1G 26380-500MG 26380-5G 26380-25G
77467-770EA 114.33 USD
77467-770 77480-472 77490-808 77500-884
Ethyl 2,6-dibromopyridine-4-carboxylate ≥97%
Ethyl 2,6-dibromopyridine-4-carboxylate

Precaución: For research use only

Formula: C₈H₇Br₂NO₂
MW: 308.955 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042544
CAS Number: 90050-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR