3-Bromo-2-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

26344-1G 26344-500MG 26344-5G 26344-25G
77467-766EA 140.08 USD
77467-766 77480-468 77490-804 77500-880
3-Bromo-2-methoxybenzyl alcohol ≥97%
3-Bromo-2-methoxybenzyl alcohol

Precaución: For research use only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238551
CAS Number: 205873-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR