1-Methylethyl 3-hydroxy-4-methylhexanoate ≥95%

Supplier: AOBChem USA

Warning

263435-1G 263435-500MG 263435-5G 263435-25G
77467-762EA 415.09 USD
77467-762 77480-464 77490-800 77500-876
1-Methylethyl 3-hydroxy-4-methylhexanoate ≥95%
1-Methylethyl 3-hydroxy-4-methylhexanoate

Precaución: For research use only

Formula: C₁₀H₂₀O₃
MW: 188.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34876915
CAS Number: 931088-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR