3-Cyclopropyl-4-fluoroaniline ≥95%

Supplier: AOBChem USA

Warning

26288-1G 26288-5G 26288-25G
77467-754EA 63.86 USD
77467-754 77490-792 77500-868
3-Cyclopropyl-4-fluoroaniline ≥95%
3-Cyclopropyl-4-fluoroaniline

Precaución: For research use only

Formula: C₉H₁₀FN
Storage Temperature: Ambient
MDL Number: MFCD19443457
CAS Number: 890129-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR