(2-Fluoro-6-(trifluoromethoxy)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

262462-1G 262462-500MG 262462-5G 262462-25G
77467-750EA 415.09 USD
77467-750 77480-454 77490-788 77500-866
(2-Fluoro-6-(trifluoromethoxy)phenyl)trimethylsilane ≥95%
(2-Fluoro-6-(trifluoromethoxy)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₂F₄OSi
MW: 252.2807 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34876650
CAS Number: 1860877-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR