1-(6-Chloro-4-methyl-2-pyridinyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

261808-1G 261808-500MG 261808-5G 261808-25G
77467-746EA 341.96 USD
77467-746 77480-450 77490-784 77500-864
1-(6-Chloro-4-methyl-2-pyridinyl)ethanone ≥95%
1-(6-Chloro-4-methyl-2-pyridinyl)ethanone

Precaución: For research use only

Formula: C₈H₈ClNO
MW: 169.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257996
CAS Number: 1256812-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR