2-Ethoxy-4,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

26166-1G 26166-500MG 26166-5G 26166-25G
77467-742EA 82.4 USD
77467-742 77480-446 77490-780 77500-860
2-Ethoxy-4,5-difluorobenzaldehyde ≥97%
2-Ethoxy-4,5-difluorobenzaldehyde

Precaución: For research use only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247322
CAS Number: 883535-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR