1-(4-Isobutoxyphenyl)propan-1-one ≥96%

Supplier: AOBChem USA

Warning

261396-1G 261396-5G 261396-25G
77467-738EA 66.95 USD
77467-738 77490-776 77500-856
1-(4-Isobutoxyphenyl)propan-1-one ≥96%
1-(4-Isobutoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD02176134
CAS Number: 354539-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR