1-isopropyl-4-methyl-1H-pyrazole ≥96%

Supplier: AOBChem USA

Warning

261351-1G 261351-500MG 261351-5G
77467-734EA 644.78 USD
77467-734 77480-442 77490-772
1-isopropyl-4-methyl-1H-pyrazole ≥96%
1-isopropyl-4-methyl-1H-pyrazole

Precaución: For research use only

Formula: C₇H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD14609314
CAS Number: 130098-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR