1-Bromo-3,5-diethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

261337-1G 261337-500MG 261337-5G 261337-25G
77467-730EA 415.09 USD
77467-730 77480-438 77490-768 77500-850
1-Bromo-3,5-diethoxybenzene ≥97%
1-Bromo-3,5-diethoxybenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO₂
MW: 245.1130 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34876464
CAS Number: 2188184-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR