[4-(Cyclopentyloxy)phenyl]boronic acid ≥96%

Supplier: AOBChem USA

Warning

Synonyms: B-[4-(Cyclopentyloxy)phenyl]boronic acid

260757-1G 260757-500MG 260757-5G 260757-25G
77467-710EA 220.42 USD
77467-710 77480-418 77490-748 77500-830
[4-(Cyclopentyloxy)phenyl]boronic acid ≥96%
[4-(Cyclopentyloxy)phenyl]boronic acid

Precaución: For research use only

Formula: C₁₁H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD08461671
CAS Number: 871830-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR