6-Bromo-2-fluoro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

26023-1G 26023-500MG 26023-5G 26023-25G
77467-702EA 149.35 USD
77467-702 77480-410 77490-738 77500-814
6-Bromo-2-fluoro-3-methylbenzyl alcohol ≥97%
6-Bromo-2-fluoro-3-methylbenzyl alcohol

Precaución: For research use only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24848714
CAS Number: 1449008-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR