4-Methoxy-3-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

26003-1G 26003-500MG 26003-5G 26003-25G
77467-698EA 426.42 USD
77467-698 77480-406 77490-734 77500-812
4-Methoxy-3-isopropylbenzoic acid ≥97%
4-Methoxy-3-isopropylbenzoic acid

Precaución: For research use only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16303871
CAS Number: 33537-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR