α-Cyclopropyl-2,6-difluoro-4-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259535-1G 259535-500MG 259535-5G
77467-694EA 597.4 USD
77467-694 77480-402 77490-730
α-Cyclopropyl-2,6-difluoro-4-methoxybenzenemethanol ≥95%
α-Cyclopropyl-2,6-difluoro-4-methoxybenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12786567
CAS Number: 1250042-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR