4-Bromo-3-chloro-α-cyclopropyl-2-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259528-1G 259528-500MG 259528-5G
77467-690EA 546.93 USD
77467-690 77480-398 77490-726
4-Bromo-3-chloro-α-cyclopropyl-2-fluorobenzenemethanol ≥95%
4-Bromo-3-chloro-α-cyclopropyl-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₁₀H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD30197492
CAS Number: 1701582-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR