α-Cyclopropyl-2,5-dimethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259513-1G 259513-500MG 259513-5G 259513-25G
77467-686EA 397.58 USD
77467-686 77480-394 77490-722 77500-810
α-Cyclopropyl-2,5-dimethoxybenzenemethanol ≥95%
α-Cyclopropyl-2,5-dimethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD11936266
CAS Number: 1181593-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR