α-Cyclopropyl-2,4,6-trifluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

259505-1G 259505-500MG 259505-5G 259505-25G
77467-682EA 312.09 USD
77467-682 77480-390 77490-718 77500-806
α-Cyclopropyl-2,4,6-trifluorobenzenemethanol ≥97%
α-Cyclopropyl-2,4,6-trifluorobenzenemethanol

Precaución: For research use only

Formula: C₁₀H₉F₃O
MW: 202.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787161
CAS Number: 1247059-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR