α-Cyclopropyl-2,6-difluoro-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259484-1G 259484-500MG 259484-5G
77467-678EA 546.93 USD
77467-678 77480-386 77490-714
α-Cyclopropyl-2,6-difluoro-3-methylbenzenemethanol ≥95%
α-Cyclopropyl-2,6-difluoro-3-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD24310357
CAS Number: 1554846-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR