α-Cyclopropyl-2-ethoxy-5-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259443-1G 259443-500MG 259443-5G
77467-674EA 477.92 USD
77467-674 77480-382 77490-710
α-Cyclopropyl-2-ethoxy-5-methylbenzenemethanol ≥95%
α-Cyclopropyl-2-ethoxy-5-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16705083
CAS Number: 1249856-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR