α-Cyclopropyl-3,5-bis(1,1-dimethylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259414-1G 259414-500MG 259414-5G 259414-25G
77467-670EA 415.09 USD
77467-670 77480-378 77490-706 77500-800
α-Cyclopropyl-3,5-bis(1,1-dimethylethyl)benzenemethanol ≥95%
α-Cyclopropyl-3,5-bis(1,1-dimethylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₈H₂₈O
Storage Temperature: Ambient
MDL Number: MFCD34875814
CAS Number: 2801693-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR