α-Cyclopropyl-5-methoxy-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259387-1G 259387-500MG 259387-5G
77467-666EA 497.49 USD
77467-666 77480-374 77490-702
α-Cyclopropyl-5-methoxy-2-methylbenzenemethanol ≥95%
α-Cyclopropyl-5-methoxy-2-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O₂
MW: 192.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34875804
CAS Number: 2294363-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR