α-Cyclopropyl-2,6-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259377-1G 259377-500MG 259377-5G
77467-662EA 434.66 USD
77467-662 77480-370 77490-698
α-Cyclopropyl-2,6-difluorobenzenemethanol ≥95%
α-Cyclopropyl-2,6-difluorobenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₀F₂O
MW: 184.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16293511
CAS Number: 1255306-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR