α-Cyclopropyl-4-propoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259373-1G 259373-500MG 259373-5G 259373-25G
77467-658EA 347.11 USD
77467-658 77480-366 77490-694 77500-796
α-Cyclopropyl-4-propoxybenzenemethanol ≥95%
α-Cyclopropyl-4-propoxybenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD17224005
CAS Number: 1341986-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR