α-Cyclopropyl-3,5-dimethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259371-1G 259371-500MG 259371-5G 259371-25G
77467-654EA 397.58 USD
77467-654 77480-362 77490-690 77500-794
α-Cyclopropyl-3,5-dimethoxybenzenemethanol ≥95%
α-Cyclopropyl-3,5-dimethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD21328335
CAS Number: 1479370-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR